Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TM6SF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144903.1 | NP_001138375.1 | 3, 4 | 411 | ||
NM_023003.3 | NP_075379.2 | 3, 4 | 411 | |||
XM_011521679.2 | XP_011519981.1 | 3, 4 | 336 | |||
XM_011521680.2 | XP_011519982.1 | 3, 4 | 336 | |||
XM_017022320.1 | XP_016877809.1 | 3, 4 | 336 | |||
XM_017022321.1 | XP_016877810.1 | 3, 4 | 336 | |||
XM_017022322.1 | XP_016877811.1 | 3, 4 | 336 | |||
XM_017022323.1 | XP_016877812.1 | 3, 4 | 463 | |||
XM_017022324.1 | XP_016877813.1 | 3, 4 | 440 | |||
XM_017022325.1 | XP_016877814.1 | 3, 4 | 272 | |||
XM_017022326.1 | XP_016877815.1 | 3, 4 | 272 | |||
XM_017022327.1 | XP_016877816.1 | 3, 4 | 464 | |||
XM_017022328.1 | XP_016877817.1 | 3, 4 | 406 | |||
XM_017022329.1 | XP_016877818.1 | 3, 4 | 396 | |||
XM_017022330.1 | XP_016877819.1 | 3, 4 | 396 | |||
XM_017022331.1 | XP_016877820.1 | 3, 4 | 396 | |||
GenBank mRNA | AF255922.1 | - | 3, 4 | 359 | ||
AK223246.1 | - | 3, 4 | 347 | |||
AK292100.1 | - | 3, 4 | 336 | |||
AK295568.1 | - | 3, 4 | 300 | |||
BC032007.1 | - | 3, 4 | 325 | |||
BC063487.1 | - | 3, 4 | 411 | 5240735 | ||
DQ021909.1 | - | 3, 4 | 302 |