Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPATS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293285.1 | NP_001280214.1 | 5 | 1156 | ||
NM_001293286.1 | NP_001280215.1 | 3 | 1009 | |||
NM_023071.3 | NP_075559.2 | 4 | 1072 | |||
NR_120663.1 | 4 | 1072 | ||||
XM_011538690.2 | XP_011536992.1 | 5 | 544 | |||
XM_011538691.2 | XP_011536993.1 | 3 | 439 | |||
XM_017019840.1 | XP_016875329.1 | 4 | 512 | |||
GenBank mRNA | AB075515.1 | - | 2 | 100 | ||
AK023202.1 | - | 4 | 471 | |||
AK095582.1 | - | 4 | 1072 | |||
AK292334.1 | - | 3 | 409 | |||
AL833614.1 | - | 5 | 597 | |||
BC048299.1 | - | 3 | 439 | 5520818 | ||
BX648916.1 | - | 3 | 381 | |||
CN421379.1 | - | 4 | 498 |