Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PYCRL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023078.3 | NP_075566.2 | 1, 2 | 165 | ||
GenBank mRNA | AK001500.2 | - | 1, 2 | 102 | ||
AK023914.1 | - | 1, 2 | 125 | |||
AK222685.1 | - | 135 | ||||
AK225404.1 | - | 1, 2 | 114 | |||
AK294615.1 | - | 2, 3 | 233 | |||
AK301225.1 | - | 1, 2 | 120 | |||
BC007993.1 | - | 1, 2 | 145 | 3533609 | ||
DA403767.1 | - | 1, 2 | 165 | |||
DA629534.1 | - | 1, 2 | 163 |