Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP4F12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023944.3 | NP_076433.3 | 4 | 487 | ||
NR_117085.1 | 4 | 411 | ||||
XM_011528203.2 | XP_011526505.1 | 1 | 26 | |||
XM_011528204.2 | XP_011526506.1 | 1 | 26 | |||
XM_011528207.2 | XP_011526509.1 | 1 | 26 | |||
XM_011528208.2 | XP_011526510.1 | 3 | 126 | |||
XM_017027172.1 | XP_016882661.1 | 4 | 429 | |||
GenBank mRNA | AB035130.1 | - | 4 | 398 | ||
AB035131.1 | - | 4 | 398 | |||
AK075435.1 | - | 4 | 339 | |||
AK304138.1 | - | 4 | 430 | |||
AK304389.1 | - | 4 | 429 | |||
AL832171.1 | - | 4 | 340 | |||
AY008841.1 | - | 3 | 368 | |||
AY358977.1 | - | 4 | 409 | |||
BC035350.1 | - | 4 | 414 | 5187264 | ||
DB221158.1 | - | 4 | 430 | |||
DC418592.1 | - | 4 | 429 |