Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHID1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142674.1 | NP_001136146.1 | 5 | 546 | ||
NM_001142675.1 | NP_001136147.1 | 4 | 656 | |||
NM_001142676.1 | NP_001136148.1 | 5 | 517 | |||
NM_001142677.1 | NP_001136149.1 | 4 | 442 | |||
NM_023947.3 | NP_076436.3 | 4 | 442 | |||
XM_005253072.2 | XP_005253129.1 | 4 | 529 | |||
XM_005253073.4 | XP_005253130.1 | 5 | 530 | |||
XM_006718282.2 | XP_006718345.1 | 5 | 517 | |||
XM_011520299.1 | XP_011518601.1 | 4 | 753 | |||
XM_011520300.1 | XP_011518602.1 | 4 | 753 | |||
XM_011520301.2 | XP_011518603.1 | 4 | 753 | |||
XM_011520302.2 | XP_011518604.1 | 4 | 753 | |||
XM_017018166.1 | XP_016873655.1 | 5 | 603 | |||
XM_017018167.1 | XP_016873656.1 | 4 | 408 | |||
XM_017018168.1 | XP_016873657.1 | 5 | 529 | |||
GenBank mRNA | AF212229.1 | - | 5 | 480 | ||
AK027711.1 | - | 4 | 391 | |||
AK075413.1 | - | 5 | 453 | |||
AK124697.1 | - | 4 | 1018 | |||
AK225025.1 | - | 5 | 482 | |||
AK225398.1 | - | 5 | 465 | |||
AK297743.1 | - | 4 | 523 | |||
AK301245.1 | - | 4 | 502 | |||
AY037151.1 | - | 4 | 403 | |||
BC000001.2 | - | 5 | 466 | 3504261 | ||
BC013642.1 | - | 2 | 78 | |||
BC095409.1 | - | 4 | 378 | 30333427 | ||
BI818660.1 | - | 5 | 546 | |||
BI821281.1 | - | 4 | 408 |