Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFNA13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006900.3 | NP_008831.3 | 1 | 691 | ||
GenBank mRNA | BC093988.1 | - | 1 | 663 | 7939833 | |
BC112002.1 | - | 1 | 663 | 8327470 |
Gene Symbol: IFNA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024013.2 | NP_076918.1 | 1 | 677 | ||
GenBank mRNA | AB578886.1 | - | 1 | 690 | ||
V00537.1 | - | 1 | 679 | |||
V00538.1 | - | 1 | 677 |