Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COLEC11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001255982.1 | NP_001242911.1 | 3 | 519 | ||
NM_001255985.1 | NP_001242914.1 | 5 | 481 | |||
NM_001255986.1 | NP_001242915.1 | 3 | 290 | |||
NM_001255987.1 | NP_001242916.1 | 2 | 218 | |||
NM_024027.4 | NP_076932.1 | 4 | 591 | |||
NM_199235.2 | NP_954705.1 | 5 | 670 | |||
NR_045659.1 | 4 | 591 | ||||
XM_005263853.4 | XP_005263910.1 | 3 | 213 | |||
GenBank mRNA | AB119525.1 | - | 4 | 477 | ||
AB119650.1 | - | 3 | 405 | |||
AB119684.1 | - | 3 | 290 | |||
AB119685.1 | - | 2 | 218 | |||
AK303824.1 | - | 5 | 481 | |||
AK313840.1 | - | 4 | 593 | |||
AY358439.1 | - | 4 | 294 | |||
BC000078.2 | - | 4 | 301 | 3507377 | ||
BC009951.2 | - | 5 | 453 | 4125795 | ||
BF316908.1 | - | 4 | 374 | |||
BM547424.1 | - | 3 | 269 |