Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GDAP1L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256737.1 | NP_001243666.1 | 6 | 1436 | ||
NM_001256738.1 | NP_001243667.1 | 6 | 1226 | |||
NM_001256739.1 | NP_001243668.1 | 5 | 1205 | |||
NM_001256740.1 | NP_001243669.1 | 4 | 1166 | |||
NM_024034.4 | NP_076939.3 | 6 | 1379 | |||
NR_046353.1 | 4 | 1012 | ||||
GenBank mRNA | AK056251.1 | - | 6 | 1369 | ||
AK293484.1 | - | 5 | 1155 | |||
AK297348.1 | - | 4 | 914 | |||
CR749676.1 | - | 2 | 667 |