Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACTR8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022899.4 | NP_075050.3 | 9 | 1192 | ||
XM_005265587.4 | XP_005265644.1 | 9 | 1156 | |||
XM_011534249.2 | XP_011532551.1 | 9 | 1160 | |||
GenBank mRNA | AK022996.1 | - | 9 | 1142 | ||
AK026232.1 | - | 4 | 345 | |||
AK094507.1 | - | 9 | 1090 | |||
BC032744.1 | - | 4 | 363 | 5575464 |