Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RALGDS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042368.2 | NP_001035827.1 | 11, 12 | 1829 | ||
NM_001271774.1 | NP_001258703.1 | 11, 12 | 1741 | |||
NM_001271775.1 | NP_001258704.1 | 11, 12 | 1834 | |||
NM_001271776.1 | NP_001258705.1 | 11, 12 | 1814 | |||
NM_006266.3 | NP_006257.1 | 11, 12 | 1850 | |||
GenBank mRNA | AB037729.1 | - | 7, 8 | 1142 | ||
AF027169.1 | - | 11, 12 | 1745 | |||
AF295773.1 | - | 11, 12 | 1765 | |||
AK000242.1 | - | 6, 7 | 807 | |||
AK024575.1 | - | 3, 4 | 404 | |||
AK056462.1 | - | 10, 11 | 1527 | |||
AK074114.1 | - | 11, 12 | 1720 | |||
AK090450.1 | - | 7, 8 | 1176 | |||
AK127524.1 | - | 11, 12 | 1834 | |||
AK301470.1 | - | 11, 12 | 1701 | |||
BC033198.1 | - | 3, 4 | 246 | |||
BC059362.1 | - | 11, 12 | 1804 | 30340922 |