Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RSRC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023012.5 | NP_075388.2 | 8 | 1024 | ||
NR_036434.1 | 9 | 1105 | ||||
NR_036435.1 | 9 | 1124 | ||||
NR_036436.1 | 8 | 1046 | ||||
XM_005253601.1 | XP_005253658.1 | 8 | 1027 | |||
XM_005253602.2 | XP_005253659.1 | 8 | 998 | |||
XM_005253604.1 | XP_005253661.1 | 9 | 1109 | |||
XM_011538688.1 | XP_011536990.1 | 8 | 1027 | |||
XM_011538689.1 | XP_011536991.1 | 11 | 1282 | |||
XM_017019830.1 | XP_016875319.1 | 8 | 1024 | |||
XM_017019831.1 | XP_016875320.1 | 9 | 1104 | |||
XM_017019832.1 | XP_016875321.1 | 11 | 1260 | |||
XM_017019833.1 | XP_016875322.1 | 9 | 1051 | |||
GenBank mRNA | AB212664.1 | - | 8 | 1010 | ||
AF161432.1 | - | 4 | 502 | |||
AK001883.1 | - | 8 | 984 | |||
AK023985.1 | - | 5 | 614 | |||
AK056114.1 | - | 9 | 1047 | |||
AK056256.1 | - | 8 | 984 | |||
BC008684.1 | - | 8 | 934 | 3853928 | ||
BC067773.1 | - | 8 | 994 | 5301415 | ||
BX640711.1 | - | 8 | 970 | |||
BX647884.1 | - | 8 | 1013 |