Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PYCRL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023078.3 | NP_075566.2 | 2 | 211 | ||
GenBank mRNA | AK001500.2 | - | 2 | 148 | ||
AK023914.1 | - | 2 | 171 | |||
AK222685.1 | - | 181 | ||||
AK225404.1 | - | 2 | 160 | |||
AK294615.1 | - | 3 | 279 | |||
AK295444.1 | - | 2 | 152 | |||
AK301225.1 | - | 2 | 166 | |||
AL833857.1 | - | 1 | 55 | |||
BC007993.1 | - | 2 | 191 | 3533609 | ||
DA403767.1 | - | 2 | 211 | |||
DA629534.1 | - | 2 | 209 |