Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MOSPD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040097.1 | NP_001035186.1 | 6 | 1061 | ||
NM_001040098.1 | NP_001035187.1 | 6 | 977 | |||
NM_001040099.1 | NP_001035188.1 | 5 | 1130 | |||
NM_023948.4 | NP_076438.1 | 5 | 1160 | |||
XM_005250529.4 | XP_005250586.1 | 6 | 982 | |||
XM_005250531.3 | XP_005250588.1 | 6 | 1007 | |||
GenBank mRNA | BC005042.1 | - | 5 | 1089 | 3689026 | |
BC011653.2 | - | 5 | 1141 | 4299899 | ||
BC028475.2 | - | 5 | 1065 |