Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDX50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024045.1 | NP_076950.1 | 2 | 213 | ||
XM_005270148.2 | XP_005270205.1 | 3 | 258 | |||
XM_011540144.2 | XP_011538446.1 | 3 | 265 | |||
XM_017016626.1 | XP_016872115.1 | 2 | 205 | |||
XM_017016627.1 | XP_016872116.1 | 4 | 681 | |||
XM_017016628.1 | XP_016872117.1 | 3 | 507 | |||
XM_017016629.1 | XP_016872118.1 | 2 | 395 | |||
XM_017016630.1 | XP_016872119.1 | 3 | 221 | |||
GenBank mRNA | AB051075.1 | - | 2 | 194 | ||
AF334103.1 | - | 2 | 190 | |||
AK223297.1 | - | 2 | 211 | |||
AK293577.1 | - | 2 | 172 | |||
AK301431.1 | - | 2 | 177 | |||
BC000272.1 | - | 2 | 213 | 3357684 |