Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C12orf43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286191.1 | NP_001273120.1 | 6 | 1544 | ||
NM_001286192.1 | NP_001273121.1 | 6 | 1454 | |||
NM_001286195.1 | NP_001273124.1 | 6 | 1421 | |||
NM_001286196.1 | NP_001273125.1 | 6 | 1418 | |||
NM_001286197.1 | NP_001273126.1 | 5 | 1355 | |||
NM_001286198.1 | NP_001273127.1 | 7 | 1711 | |||
NM_022895.2 | NP_075046.1 | 6 | 1451 | |||
NR_104409.1 | 6 | 1464 | ||||
XM_017019829.1 | XP_016875318.1 | 6 | 1535 | |||
GenBank mRNA | AK022510.1 | - | 6 | 1446 | ||
BC014661.1 | - | 6 | 1428 | 4860511 |