Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC26A6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040454.1 | NP_001035544.1 | 20 | 2717 | ||
NM_001281732.1 | NP_001268661.1 | 20 | 2521 | |||
NM_001281733.1 | NP_001268662.1 | 19 | 2305 | |||
NM_022911.2 | NP_075062.2 | 21 | 2629 | |||
NM_134263.2 | NP_599025.2 | 21 | 2626 | |||
NM_134426.2 | NP_602298.2 | 20 | 2572 | |||
GenBank mRNA | AB033288.1 | - | 20 | 2535 | ||
AB102713.1 | - | 20 | 2617 | |||
AB208936.1 | - | 12 | 5670 | |||
AF279265.1 | - | 20 | 2717 | |||
AF288410.1 | - | 21 | 2577 | |||
AF416721.1 | - | 21 | 2574 | |||
AK056237.1 | - | 19 | 2305 | |||
AL050170.1 | - | 2 | 416 | |||
BC017697.1 | - | 21 | 2575 | 4398446 |