Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TFE3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282142.1 | NP_001269071.1 | 10 | 3090 | ||
NM_006521.5 | NP_006512.2 | 10 | 3155 | |||
GenBank mRNA | AL161985.1 | - | 10 | 2991 | ||
BC026027.1 | - | 10 | 3102 | 4576858 | ||
X51330.1 | - | 7 | 2236 | |||
X96717.1 | - | 10 | 3101 |