Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COA7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023077.2 | NP_075565.2 | 3 | 1113 | ||
GenBank mRNA | AK022501.1 | - | 3 | 1113 | ||
AK023237.1 | - | 1 | 398 | |||
AK225041.1 | - | 3 | 1088 | |||
AK225087.1 | - | 3 | 1094 |