Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHID1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142674.1 | NP_001136146.1 | 11, 12 | 1214 | ||
NM_001142675.1 | NP_001136147.1 | 10, 11 | 1324 | |||
NM_001142676.1 | NP_001136148.1 | 11, 12 | 1185 | |||
NM_001142677.1 | NP_001136149.1 | 9, 10 | 1017 | |||
NM_023947.3 | NP_076436.3 | 10, 11 | 1110 | |||
XM_005253072.2 | XP_005253129.1 | 10, 11 | 1197 | |||
XM_005253073.4 | XP_005253130.1 | 10, 11 | 1105 | |||
XM_006718282.2 | XP_006718345.1 | 11, 12 | 1185 | |||
XM_011520299.1 | XP_011518601.1 | 10, 11 | 1421 | |||
XM_011520300.1 | XP_011518602.1 | 9, 10 | 1328 | |||
XM_017018166.1 | XP_016873655.1 | 11, 12 | 1271 | |||
GenBank mRNA | AF212229.1 | - | 11, 12 | 1148 | ||
AK027711.1 | - | 10, 11 | 1059 | |||
AK075413.1 | - | 11, 12 | 1121 | |||
AK124697.1 | - | 10, 11 | 1686 | |||
AK225025.1 | - | 11, 12 | 1150 | |||
AK225398.1 | - | 11, 12 | 1133 | |||
AK297743.1 | - | 10, 11 | 1191 | |||
AK301245.1 | - | 10, 11 | 1170 | |||
AY037151.1 | - | 9, 10 | 978 | |||
BC000001.2 | - | 11, 12 | 1134 | 3504261 | ||
BC013642.1 | - | 8, 9 | 746 | |||
BC095409.1 | - | 10, 11 | 1046 | 30333427 |