Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPATA5L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001323640.1 | NP_001310569.1 | 3 | 1398 | ||
NM_024063.2 | NP_076968.2 | 3 | 1398 | |||
NR_027635.1 | 3 | 1398 | ||||
NR_136645.1 | 3 | 1405 | ||||
NR_136646.1 | 4 | 1593 | ||||
NR_136647.1 | 4 | 1560 | ||||
NR_136648.1 | 4 | 1593 | ||||
GenBank mRNA | AK023232.1 | - | 3 | 1344 | ||
AK225463.1 | - | 3 | 1344 | |||
AK291457.1 | - | 3 | 1351 | |||
BC000981.2 | - | 3 | 1331 | 2985725 |