Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRRG4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024081.5 | NP_076986.1 | 2 | 339 | ||
XM_006718313.3 | XP_006718376.3 | 2 | 518 | |||
XM_006718314.3 | XP_006718377.1 | 2 | 189 | |||
XM_017018275.1 | XP_016873764.1 | 2 | 499 | |||
GenBank mRNA | AF326351.1 | - | 2 | 187 | ||
BC010052.2 | - | 2 | 339 | 3841745 | ||
BC063393.1 | - | 2 | 331 | 5183337 | ||
KU178766.1 | - | 1 | 78 | |||
KU178767.1 | - | 1 | 78 | |||
KU178768.1 | - | 1 | 78 |