Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DSCC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024094.2 | NP_076999.2 | 1, 2 | 314 | ||
XM_005251065.3 | XP_005251122.1 | 1, 2 | 305 | |||
GenBank mRNA | AK054585.1 | - | 1, 2 | 286 | ||
AK074355.1 | - | 1, 2 | 252 | |||
AK315356.1 | - | 1, 2 | 241 | |||
AY358866.1 | - | 1, 2 | 266 | |||
BC001316.1 | - | 1, 2 | 293 | 3454505 | ||
BC001531.2 | - | 1, 2 | 252 | 3510131 | ||
DA346021.1 | - | 1, 2 | 315 |