Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC440040 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027044.1 | 7 | 1744 | |||
GenBank mRNA | AK294678.1 | - | 7 | 1744 | ||
BC141819.1 | - | 7 | 1652 | |||
BC142657.1 | - | 7 | 1652 |
Gene Symbol: TRIM48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024114.3 | NP_077019.2 | 6 | 1001 | ||
GenBank mRNA | AF521869.1 | - | 6 | 1008 | ||
BC001862.2 | - | 6 | 1001 | 3603763 |