Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUP98 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | KU847959.1 | - | 1934 |
Gene Symbol: PHF23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284517.1 | NP_001271446.1 | 4 | 577 | ||
NM_001284518.1 | NP_001271447.1 | 4 | 662 | |||
NM_024297.2 | NP_077273.2 | 4 | 778 | |||
GenBank mRNA | AK026537.1 | - | 4 | 722 | ||
AK074766.1 | - | 4 | 721 | |||
AK122791.1 | - | 3 | 695 | |||
AK122901.1 | - | 4 | 662 | |||
AK297040.1 | - | 4 | 510 | |||
BC002509.2 | - | 4 | 726 | 3051214 | ||
BC008630.1 | - | 1 | 288 | |||
EF071959.1 | - | 4 | 724 |