Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THTPA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001126339.3 | NP_001119811.1 | 2 | 625 | ||
NM_001256062.2 | NP_001242991.1 | 1 | 1024 | |||
NM_001256321.2 | NP_001243250.1 | 2 | 625 | |||
NM_001256322.2 | NP_001243251.1 | 1 | 1024 | |||
NM_001256323.2 | NP_001243252.1 | 2 | 625 | |||
NM_024328.5 | NP_077304.1 | 1 | 1024 | |||
GenBank mRNA | AF432862.1 | - | 1 | 293 | ||
AK057691.1 | - | 1 | 731 | |||
AK098604.1 | - | 2 | 414 | |||
BC002984.1 | - | 1 | 718 | 3535451 | ||
BI522644.1 | - | 2 | 429 | |||
BM925597.1 | - | 2 | 612 | |||
BP358780.1 | - | 2 | 569 | |||
BX161435.1 | - | 1 | 984 | |||
BX378775.2 | - | 1 | 712 | |||
DA952722.1 | - | 2 | 339 | |||
DB122722.1 | - | 2 | 336 |