Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDCL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024065.4 | NP_076970.1 | 3 | 291 | ||
XM_017004890.1 | XP_016860379.1 | 3 | 420 | |||
GenBank mRNA | AF110511.1 | - | 3 | 240 | ||
AF267853.1 | - | 3 | 243 | |||
AK313985.1 | - | 3 | 557 | |||
BC001021.2 | - | 3 | 270 | 3344703 | ||
BG819099.1 | - | 3 | 282 | |||
BI820064.1 | - | 3 | 292 |
Gene Symbol: PDCL3P4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002941.2 | 1 | 218 | |||
GenBank mRNA | AK022267.1 | - | 1 | 1716 |