Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZFYVE21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198953.1 | NP_001185882.1 | 7, 8 | 829 | ||
NM_024071.3 | NP_076976.1 | 6, 7 | 775 | |||
GenBank mRNA | AK001921.1 | - | 1, 2 | 1340 | ||
AK057816.1 | - | 7, 8 | 750 | |||
AK290519.1 | - | 6, 7 | 688 | |||
AY395756.1 | - | 6, 7 | 690 | |||
BC001130.2 | - | 6, 7 | 696 | 2966683 | ||
BC005999.1 | - | 6, 7 | 711 | 4091809 | ||
BX248261.1 | - | 6, 7 | 681 | |||
CX786145.1 | - | 7, 8 | 829 |