Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATG9A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077198.2 | NP_001070666.1 | 11 | 1890 | ||
NM_024085.4 | NP_076990.4 | 10 | 1838 | |||
NR_104255.1 | 11 | 1831 | ||||
GenBank mRNA | AK025822.1 | - | 5 | 1192 | ||
AK056595.1 | - | 11 | 1831 | |||
AK289755.1 | - | 11 | 1785 | |||
AK302521.1 | - | 10 | 1729 | |||
AK315553.1 | - | 5 | 1202 | |||
AL833865.1 | - | 4 | 1087 | |||
BC001098.2 | - | 3 | 365 | |||
BC021202.2 | - | 2 | 196 | |||
BC065534.1 | - | 11 | 1769 | 5787445 | ||
BX537984.1 | - | 10 | 1759 | |||
BX538192.1 | - | 10 | 1733 | |||
BX538198.1 | - | 11 | 1667 |