Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCTPP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024096.1 | NP_077001.1 | 3 | 908 | ||
NR_134470.1 | 3 | 933 | ||||
NR_134471.1 | 3 | 947 | ||||
GenBank mRNA | AF210430.1 | - | 3 | 881 | ||
AF212242.1 | - | 3 | 879 | |||
AK024843.1 | - | 3 | 876 | |||
AY453409.1 | - | 3 | 908 | |||
BC001344.1 | - | 3 | 908 | 3463032 |