Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR77 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317062.1 | NP_001303991.1 | 4 | 618 | ||
NM_001317063.1 | NP_001303992.1 | 4 | 618 | |||
NM_024102.3 | NP_077007.1 | 4 | 618 | |||
NR_133654.1 | 3 | 476 | ||||
GenBank mRNA | AB073603.1 | - | 4 | 566 | ||
AF052183.1 | - | 4 | 504 | |||
AF478464.1 | - | 4 | 479 | |||
AK022860.1 | - | 4 | 548 | |||
AK223189.1 | - | 4 | 548 | |||
AK296163.1 | - | 3 | 420 | |||
AL557974.3 | - | 4 | 534 | |||
AY225316.1 | - | 4 | 504 | |||
BC001679.2 | - | 4 | 539 | 2822173 | ||
BC006477.1 | - | 4 | 539 | 2822173 | ||
BC009411.1 | - | 4 | 551 | 3614223 | ||
BC011778.2 | - | 4 | 558 | 3941878 | ||
BC016946.1 | - | 4 | 518 | 3845908 | ||
BI195029.1 | - | 3 | 290 |