Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF426 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318056.1 | NP_001304985.1 | 4, 5 | 407 | ||
NM_024106.2 | NP_077011.1 | 4, 5 | 407 | |||
GenBank mRNA | AK095759.1 | - | 2, 3 | 462 | ||
BC001791.1 | - | 4, 5 | 389 | 3543910 | ||
BE315219.1 | - | 4, 5 | 352 |