Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1orf116 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083924.1 | NP_001077393.1 | 3 | 3547 | ||
NM_023938.5 | NP_076427.2 | 4 | 3733 | |||
XM_005273259.1 | XP_005273316.1 | 2 | 3301 | |||
XM_006711530.1 | XP_006711593.1 | 4 | 3610 | |||
XM_011509973.2 | XP_011508275.1 | 3 | 3403 | |||
GenBank mRNA | AK096741.1 | - | 1 | 1349 | ||
BC001943.1 | - | 1 | 231 | |||
BC002325.2 | - | 1 | 231 |