Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFAF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039375.2 | NP_001034464.1 | 7, 8 | 745 | ||
NM_024120.4 | NP_077025.2 | 8, 9 | 829 | |||
NR_029377.1 | 9, 10 | 927 | ||||
XM_006723622.2 | XP_006723685.1 | 7, 8 | 710 | |||
XM_006723623.2 | XP_006723686.1 | 7, 8 | 842 | |||
XM_006723624.2 | XP_006723687.1 | 7, 8 | 856 | |||
XM_011529344.2 | XP_011527646.1 | 5, 6 | 1184 | |||
XM_017028061.1 | XP_016883550.1 | 8, 9 | 941 | |||
GenBank mRNA | AK025977.1 | - | 2, 3 | 255 | ||
AK091060.1 | - | 9, 10 | 904 | |||
AK289781.1 | - | 8, 9 | 821 | |||
BC005984.1 | - | 8, 9 | 639 | |||
BC073158.1 | - | 7, 8 | 722 | 6053315 |