Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHCHD7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011667.2 | NP_001011667.1 | 4 | 523 | ||
NM_001011668.2 | NP_001011668.1 | 4 | 479 | |||
NM_001011669.2 | NP_001011669.1 | 4 | 503 | |||
NM_001011670.2 | NP_001011670.1 | 3 | 375 | |||
NM_001011671.2 | NP_001011671.1 | 3 | 331 | |||
NM_001317858.1 | NP_001304787.1 | 4 | 503 | |||
NM_001317859.1 | NP_001304788.1 | 3 | 331 | |||
NM_024300.4 | NP_077276.2 | 4 | 459 | |||
NR_133934.1 | 4 | 523 | ||||
NR_133935.1 | 4 | 523 | ||||
NR_133936.1 | 4 | 479 | ||||
NR_133937.1 | 3 | 371 | ||||
GenBank mRNA | AK095922.1 | - | 3 | 180 | ||
AK098285.1 | - | 2 | 2982 | |||
AK225982.1 | - | 4 | 318 | |||
AK290088.1 | - | 4 | 308 | |||
AY070434.1 | - | 4 | 351 | |||
BC002546.2 | - | 4 | 298 | 3139026 | ||
BI911444.1 | - | 4 | 334 | |||
BM695222.1 | - | 3 | 215 | |||
BX473986.1 | - | 4 | 361 | |||
CN256918.1 | - | 3 | 161 | |||
DA367603.1 | - | 4 | 372 | |||
DA431755.1 | - | 4 | 372 | |||
DA988698.1 | - | 4 | 328 | |||
HY130470.1 | - | 4 | 460 |