Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C19orf57 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024323.3 | NP_077299.3 | 8 | 1971 | ||
XM_006722885.3 | XP_006722948.2 | 8 | 2344 | |||
XM_017027300.1 | XP_016882789.1 | 8 | 2224 | |||
XM_017027301.1 | XP_016882790.1 | 7 | 1995 | |||
GenBank mRNA | AK097431.1 | - | 8 | 1971 | ||
BC002891.2 | - | 4 | 1190 | |||
BC012945.1 | - | 5 | 1640 | |||
BC119719.1 | - | 7 | 1939 | 40117748 |