Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FOXF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001451.2 | NP_001442.2 | 2 | 1077 | ||
GenBank mRNA | AK314167.1 | - | 2 | 1084 | ||
BC089442.1 | - | 2 | 1066 | 30558276 | ||
U13219.1 | - | 2 | 1052 |