Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFS7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024407.4 | NP_077718.3 | 7 | 523 | ||
XM_005259556.4 | XP_005259613.2 | 7 | 507 | |||
GenBank mRNA | AF060512.1 | - | 7 | 493 | ||
AF115969.1 | - | 7 | 506 | |||
AK091024.1 | - | 6 | 732 | |||
AK091046.1 | - | 1 | 1425 | |||
AK091623.1 | - | 7 | 503 | |||
AK092169.1 | - | 6 | 771 | |||
AK127759.1 | - | 6 | 2433 | |||
AK128713.1 | - | 5 | 1038 | |||
AK222738.1 | - | 7 | 500 | |||
AK289671.1 | - | 8 | 781 | |||
AK293917.1 | - | 7 | 507 | |||
AK297611.1 | - | 6 | 575 | |||
BC001715.3 | - | 6 | 466 | |||
BC005954.1 | - | 7 | 500 | 4276489 | ||
BC111517.1 | - | 7 | 510 | 40017044 | ||
DC340464.1 | - | 7 | 523 |