Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NOTCH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001200001.1 | NP_001186930.1 | 5 | 1081 | ||
NM_024408.3 | NP_077719.2 | 5 | 1081 | |||
XM_005270901.3 | XP_005270958.1 | 6 | 3395 | |||
XM_011541519.2 | XP_011539821.1 | 5 | 6189 | |||
XM_011541520.2 | XP_011539822.1 | 5 | 3099 | |||
XM_017001372.1 | XP_016856861.1 | 5 | 751 | |||
XM_017001373.1 | XP_016856862.1 | 5 | 730 | |||
GenBank mRNA | AF308601.1 | - | 5 | 796 | ||
AF315356.1 | - | 5 | 796 | |||
AK307843.1 | - | 4 | 923 | |||
BC071562.1 | - | 5 | 1030 | 4822955 | ||
GQ231534.1 | - | 5 | 943 |