Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM106C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143841.1 | NP_001137313.1 | 8 | 1269 | ||
NM_001143842.1 | NP_001137314.1 | 8 | 1397 | |||
NM_001143843.1 | NP_001137315.1 | 8 | 1340 | |||
NM_024056.3 | NP_076961.1 | 8 | 1326 | |||
GenBank mRNA | BC000854.2 | - | 7 | 1239 | 3459189 | |
BC107792.1 | - | 8 | 1212 | 30321820 |