Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SECISBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282688.1 | NP_001269617.1 | 17 | 2875 | ||
NM_001282689.1 | NP_001269618.1 | 17 | 2757 | |||
NM_001282690.1 | NP_001269619.1 | 17 | 2955 | |||
NM_024077.4 | NP_076982.3 | 17 | 2878 | |||
XM_005252196.2 | XP_005252253.1 | 17 | 2728 | |||
XM_005252202.3 | XP_005252259.1 | 13 | 2110 | |||
XM_006717282.2 | XP_006717345.1 | 17 | 2811 | |||
XM_011519001.1 | XP_011517303.1 | 16 | 2842 | |||
XM_011519002.1 | XP_011517304.1 | 17 | 2818 | |||
XM_011519003.1 | XP_011517305.1 | 17 | 2815 | |||
XM_017015122.1 | XP_016870611.1 | 16 | 2755 | |||
XM_017015123.1 | XP_016870612.1 | 17 | 2731 | |||
XM_017015124.1 | XP_016870613.1 | 16 | 2638 | |||
XM_017015125.1 | XP_016870614.1 | 15 | 2433 | |||
GenBank mRNA | AB208940.1 | - | 17 | 2798 | ||
AF380995.1 | - | 17 | 2831 | |||
AK023078.1 | - | 10 | 1609 | |||
AK299161.1 | - | 17 | 2966 | |||
AK302852.1 | - | 17 | 2873 | |||
AL136881.1 | - | 17 | 2711 | |||
BC068466.1 | - | 17 | 2813 |