Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C2orf49 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286537.1 | NP_001273466.1 | 5 | 782 | ||
NM_024093.2 | NP_076998.1 | 4 | 908 | |||
XM_017004892.1 | XP_016860381.1 | 5 | 884 | |||
GenBank mRNA | AK127661.1 | - | 4 | 713 | ||
AK304268.1 | - | 5 | 782 | |||
AK307891.1 | - | 5 | 608 | |||
BC001310.1 | - | 4 | 711 | 3453623 |