Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR77 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317062.1 | NP_001303991.1 | 5, 6 | 711 | ||
NM_001317063.1 | NP_001303992.1 | 5, 6 | 711 | |||
NM_001317064.1 | NP_001303993.1 | 3, 4 | 519 | |||
NM_024102.3 | NP_077007.1 | 5, 6 | 711 | |||
NR_133654.1 | 4, 5 | 569 | ||||
GenBank mRNA | AB073603.1 | - | 5, 6 | 659 | ||
AB074171.1 | - | 1 | 23 | |||
AF052183.1 | - | 5, 6 | 597 | |||
AF478464.1 | - | 5, 6 | 572 | |||
AK022860.1 | - | 5, 6 | 641 | |||
AK223189.1 | - | 5, 6 | 641 | |||
AK298179.1 | - | 3, 4 | 463 | |||
AL557974.3 | - | 5, 6 | 627 | |||
AY225316.1 | - | 5, 6 | 597 | |||
BC001679.2 | - | 5, 6 | 632 | 2822173 | ||
BC006477.1 | - | 5, 6 | 632 | 2822173 | ||
BC009411.1 | - | 5, 6 | 644 | 3614223 | ||
BC011778.2 | - | 5, 6 | 651 | 3941878 | ||
BC016946.1 | - | 5, 6 | 611 | 3845908 | ||
BI195029.1 | - | 4, 5 | 383 |