Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHF23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284517.1 | NP_001271446.1 | 5 | 1368 | ||
NM_001284518.1 | NP_001271447.1 | 5 | 1453 | |||
NM_024297.2 | NP_077273.2 | 5 | 1569 | |||
GenBank mRNA | AK026537.1 | - | 5 | 1512 | ||
AK074766.1 | - | 5 | 1512 | |||
AK122791.1 | - | 4 | 1485 | |||
AK122901.1 | - | 5 | 1453 | |||
BC002509.2 | - | 5 | 1517 | 3051214 | ||
BC008630.1 | - | 2 | 1079 | |||
EF071959.1 | - | 5 | 1515 |