Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THTPA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001126339.3 | NP_001119811.1 | 3 | 1656 | ||
NM_001256062.2 | NP_001242991.1 | 2 | 1790 | |||
NM_001256321.2 | NP_001243250.1 | 3 | 1391 | |||
NM_001256322.2 | NP_001243251.1 | 2 | 1785 | |||
NM_001256323.2 | NP_001243252.1 | 3 | 1386 | |||
NM_024328.5 | NP_077304.1 | 2 | 2055 | |||
NR_023314.4 | 2 | 1094 | ||||
NR_046051.1 | 5 | 1197 | ||||
NR_046052.1 | 3 | 977 | ||||
GenBank mRNA | AK057691.1 | - | 2 | 1762 | ||
AK094211.1 | - | 3 | 970 | |||
AK098604.1 | - | 3 | 1445 | |||
BC002984.1 | - | 2 | 1749 | 3535451 | ||
BU535676.1 | - | 1 | 545 | |||
BX161435.1 | - | 2 | 2015 |