Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EFHD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024329.5 | NP_077305.2 | 4 | 2159 | ||
XM_005246000.3 | XP_005246057.1 | 3 | 2022 | |||
GenBank mRNA | BC003033.1 | - | 1 | 783 | ||
BC007233.1 | - | 4 | 2117 | 2961350 | ||
BC013648.1 | - | 1 | 753 | |||
BC014923.2 | - | 4 | 2113 | 3909362 | ||
BC023611.2 | - | 4 | 2109 | 4638660 | ||
BC068473.1 | - | 4 | 2129 | 5267148 |