Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM65A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193522.1 | NP_001180451.1 | 13 | 1789 | ||
NM_001193523.1 | NP_001180452.1 | 13 | 1902 | |||
NM_001193524.1 | NP_001180453.1 | 13 | 1815 | |||
NM_024519.3 | NP_078795.2 | 13 | 1780 | |||
XM_011523321.1 | XP_011521623.1 | 13 | 2594 | |||
XM_011523322.1 | XP_011521624.1 | 13 | 1789 | |||
XM_011523324.2 | XP_011521626.1 | 13 | 1849 | |||
XM_011523325.1 | XP_011521627.1 | 13 | 1804 | |||
XM_011523326.1 | XP_011521628.1 | 13 | 2003 | |||
XM_017023663.1 | XP_016879152.1 | 13 | 1789 | |||
XM_017023664.1 | XP_016879153.1 | 12 | 1675 | |||
GenBank mRNA | AB067517.1 | - | 12 | 1639 | ||
AK023787.1 | - | 8 | 1272 | |||
AK097087.1 | - | 12 | 2030 | |||
AK127792.1 | - | 13 | 1789 | |||
AK293748.1 | - | 13 | 1812 | |||
AK295677.1 | - | 13 | 1746 | |||
AK302868.1 | - | 3 | 939 | |||
AK307467.1 | - | 13 | 1733 | |||
AL834312.1 | - | 3 | 845 | |||
BC054512.1 | - | 3 | 822 | |||
BC098587.1 | - | 7 | 1217 |