Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C2orf47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024520.2 | NP_078796.2 | 2, 3 | 780 | ||
XM_005246852.2 | XP_005246909.1 | 1, 2 | 691 | |||
GenBank mRNA | AK026208.1 | - | 2, 3 | 780 | ||
AL832964.1 | - | 1, 2 | 99 | |||
BC017959.1 | - | 1, 2 | 464 | 4607945 |