Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC52A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253815.1 | NP_001240744.1 | 3 | 511 | ||
NM_001253816.1 | NP_001240745.1 | 3 | 491 | |||
NM_024531.4 | NP_078807.1 | 3 | 605 | |||
NR_045600.1 | 3 | 819 | ||||
XM_006716658.2 | XP_006716721.1 | 3 | 922 | |||
XM_006716659.2 | XP_006716722.1 | 3 | 737 | |||
XM_006716660.2 | XP_006716723.1 | 3 | 888 | |||
XM_011517300.2 | XP_011515602.1 | 2 | 282 | |||
XM_017013819.1 | XP_016869308.1 | 3 | 717 | |||
XM_017013820.1 | XP_016869309.1 | 3 | 452 | |||
GenBank mRNA | AB522904.1 | - | 2 | 394 | ||
AK021918.1 | - | 3 | 775 | |||
AK027888.1 | - | 3 | 484 | |||
AK291581.1 | - | 3 | 511 | |||
AK303822.1 | - | 3 | 410 | |||
AL560856.3 | - | 3 | 630 | |||
AY070774.1 | - | 2 | 327 | |||
BC002917.2 | - | 3 | 557 | 3954480 | ||
CN260061.1 | - | 1 | 173 | |||
DC365525.1 | - | 3 | 491 |
Set Membership: |
G-protein coupled receptor |