Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ELK4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001973.3 | NP_001964.2 | 3 | 1557 | ||
NM_021795.2 | NP_068567.1 | 3 | 1557 | |||
XM_005244950.4 | XP_005245007.1 | 4 | 467 | |||
XM_005244951.4 | XP_005245008.1 | 3 | 1261 | |||
XM_017000580.1 | XP_016856069.1 | 3 | 1711 | |||
GenBank mRNA | BC040145.1 | - | 3 | 578 | ||
BC063676.1 | - | 2 | 337 | 4364006 | ||
M85164.1 | - | 3 | 456 | |||
M85165.1 | - | 3 | 456 |