Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACSS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024560.2 | NP_078836.1 | 1, 2 | 407 | ||
XM_005269151.4 | XP_005269208.1 | 1, 2 | 419 | |||
GenBank mRNA | AK025616.1 | - | 1, 2 | 358 | ||
BC009317.2 | - | 1, 2 | 407 | 4120563 | ||
BC015769.1 | - | 1, 2 | 407 | 4902942 |